Assay Details
| Gene Symbol: LINC00330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038433.2 | 2 - 3 | 532 | 102 | ||
| GenBank mRNA | AK056732.1 | - | 2 - 3 | 533 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000624.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820720 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820759 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268067 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820758 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268069 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820765 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268155 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16631784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820672 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268161 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820699 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820773 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268089 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820757 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820711 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820748 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268074 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820753 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16631787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268066 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820806 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820744 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820703 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16631783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29268170 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08820729 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 958737 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 880972 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 502803 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 356180 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3255587 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43027 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3255587 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3255587 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11721338 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02768505 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P11721338 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1564263_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007099.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007099.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007099.hg.1 | TC13000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007097.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007097.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007097.hg.1 | TC13000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007098.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007098.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13007098.hg.1 | TC13000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008802.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008802.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008802.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008802.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008802.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008802.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159338.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159338.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159338.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159338.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159338.hg.1 | TC1300008802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008802.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008802.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008802.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008802.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008802.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008802.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159336.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159336.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159336.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159336.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159336.hg.1 | TC1300008802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300159337.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159337.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159337.hg.1 | ENST00000414562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300159337.hg.1 | NR_038433.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300159337.hg.1 | TC1300008802.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1564263_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564263_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564263_at10 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049515 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049515 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 10713472 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1754032 | AK056732.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1754032 | NR_038433.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754032 | AK056732.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754032 | NR_038433.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1754032 | AK056732.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1754032 | NR_038433.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754032 | AK056732.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754032 | NR_038433.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754032 | AK056732.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754032 | NR_038433.1 | human_transcriptome |

LINC00330
Genomic Map