Assay Details
| Gene Symbol: LOC101928314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125895.1 | 3 - 4 | 333 | 72 | ||
| GenBank mRNA | BC038759.2 | - | 2 - 3 | 304 | 72 | |
| BI461914.1 | - | 3 - 4 | 338 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928314
Genomic Map