Assay Details
| Gene Symbol: C16orf90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080524.1 | NP_001073993.1 | 1 - 2 | 41 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000818.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30404934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30404941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30404959 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894369 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30404932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894358 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894359 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30404939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894372 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09894361 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295690 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1484 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1485 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20376 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P12371257 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03104657 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3295690 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3295690 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894372 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894376 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894378 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894359 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894372 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404934 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30404941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894381 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894376 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09894382 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12371257 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894358 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894372 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894383 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894396 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16845339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894355 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894379 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894365 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894392 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894359 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894369 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894357 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894368 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894370 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894402 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16845335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894398 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16845328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894394 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09894362 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16845337 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11734236_at | - | - |
| Affymetrix Human Genome U219 Array | 11734236_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011823.hg.1 | ENST00000437192.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011823.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011823.hg.1 | TC16000818.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011822.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011822.hg.1 | TC16000818.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011825.hg.1 | ENST00000437192.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011825.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011825.hg.1 | TC16000818.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011824.hg.1 | ENST00000437192.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011824.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011824.hg.1 | TC16000818.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600170178.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170178.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170178.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170178.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170178.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170178.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170178.hg.1 | TC1600009196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170180.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170180.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170180.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170180.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170180.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170180.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170180.hg.1 | TC1600009196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600170179.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170179.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170179.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170179.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170179.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170179.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170179.hg.1 | TC1600009196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000399645.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009196.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170181.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170181.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170181.hg.1 | ENST00000437192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600170181.hg.1 | NM_001080524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600170181.hg.1 | TC1600009196.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053429 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053429 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 153646 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 180642 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1713956 | NM_001080524.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1874870 | NM_001080524.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1874870 | NM_001080524.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1713956 | NM_001080524.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1713956 | NM_001080524.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1713956 | NM_001080524.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1874870 | NM_001080524.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1713956 | NM_001080524.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1874870 | NM_001080524.1 | human_transcriptome |

C16orf90
Genomic Map