Assay Details
Gene Symbol: CCNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287253.1 | NP_001274182.1 | 1 - 2 | 484 | 109 | |
XM_017009725.1 | XP_016865214.1 | 1 - 2 | 527 | 109 | ||
GenBank mRNA | BC144416.1 | - | 1 - 2 | 466 | 9052937 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159334.hg.1 | TC0500008646.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159331.hg.1 | TC0500008646.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159332.hg.1 | TC0500008646.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000378731.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000468733.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000492179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | ENST00000614847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001039780.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287252.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008646.hg.1 | NM_001287253.1 | Refseq |
