Assay Details
| Gene Symbol: JAKMIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105521.3 | NP_001098991.1 | 5 - 6 | 1207 | 117 | |
| NM_001323086.1 | NP_001310015.1 | 5 - 6 | 1207 | 117 | ||
| NM_001323087.1 | NP_001310016.1 | 5 - 6 | 1207 | 117 | ||
| NM_001323088.1 | NP_001310017.1 | 5 - 6 | 1201 | 117 | ||
| NM_001323089.1 | NP_001310018.1 | 4 - 5 | 991 | 117 | ||
| NM_001323090.1 | NP_001310019.1 | 5 - 6 | 1207 | 117 | ||
| XM_005252674.4 | XP_005252731.1 | 5 - 6 | 3201 | 117 | ||
| XM_005252676.4 | XP_005252733.1 | 4 - 5 | 2979 | 117 | ||
| XM_011539676.2 | XP_011537978.1 | 5 - 6 | 1453 | 117 | ||
| XM_011539682.2 | XP_011537984.1 | 4 - 5 | 2985 | 117 | ||
| XM_017016095.1 | XP_016871584.1 | 5 - 6 | 3201 | 117 | ||
| XM_017016096.1 | XP_016871585.1 | 5 - 6 | 1452 | 117 | ||
| XM_017016097.1 | XP_016871586.1 | 4 - 5 | 2979 | 117 | ||
| XM_017016098.1 | XP_016871587.1 | 5 - 6 | 3201 | 117 | ||
| XM_017016099.1 | XP_016871588.1 | 5 - 6 | 3201 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000940.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10011511.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011511.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011511.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011506.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011506.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011506.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011507.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011507.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011507.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011513.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011513.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011513.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011508.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011508.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011508.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011510.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011510.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011510.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011512.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011512.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011512.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011509.hg.1 | ENST00000298622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011509.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011509.hg.1 | TC10000940.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | ENST00000477275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009396.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164170.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164170.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164170.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164170.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164170.hg.1 | TC1000009396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164165.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164165.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164165.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164165.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164165.hg.1 | TC1000009396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164172.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164172.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164172.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164172.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164172.hg.1 | TC1000009396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164166.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164166.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164166.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164166.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164166.hg.1 | TC1000009396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164167.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164167.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164167.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164167.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164167.hg.1 | TC1000009396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164171.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164171.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164171.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164171.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164171.hg.1 | TC1000009396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164169.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164169.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164169.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164169.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164169.hg.1 | TC1000009396.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164168.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164168.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164168.hg.1 | ENST00000298622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164168.hg.1 | NM_001105521.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164168.hg.1 | TC1000009396.hg.1 | Clariom D TC |

JAKMIP3
Genomic Map