Assay Details
| Gene Symbol: LINC00940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036546.1 | 2 | 387 | 115 | ||
| GenBank mRNA | AK296286.1 | - | 2 | 345 | 115 | |
| AK316036.1 | - | 2 | 385 | 115 | ||
| DA269488.1 | - | 2 | 387 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296286.1, AK316036.1 transcripts.
Set Membership: |
|

LINC00940
Genomic Map