Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000517733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000517733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000517733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000518902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000518902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
NR_108027.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
NR_108027.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013404.hg.1 |
NR_108027.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000517733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000517733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000517733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000518902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000518902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
ENST00000521128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
NR_108027.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
NR_108027.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013404.hg.1 |
NR_108027.1 |
Refseq |