Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000333503.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000562001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000562001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
ENST00000562001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011463.hg.1 |
NM_001042371.2 |
Refseq |