Assay Details
Gene Symbol: LOC399815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC053499.1 | - | 5 - 6 | 850 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC053499.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
