Assay Details
| Gene Symbol: HSP90B3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY956769.1 | - | 3 | 451 | 150 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cell division cycle 7 | CDC7 |
CDC7L1, HsCDC7, Hsk1, huCDC7 |
-117.0kb | Sense | Chr. 1 - 91500814 - 91525764 on NCBI Build GRCh38 |
| Downstream | transforming growth factor beta receptor 3 | TGFBR3 |
BGCAN, betaglycan |
36.6kb | Anti-sense | Chr. 1 - 91680343 - 91906002 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374666 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374662 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404303 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374745 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374734 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374741 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404245 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374672 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404200 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374735 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374701 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404204 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374643 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404265 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374686 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404278 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374725 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374752 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404205 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374730 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374692 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374697 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374748 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15085132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374732 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374676 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374670 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374656 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404259 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374681 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374684 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404203 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374705 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374674 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15085148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374728 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404280 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404239 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404257 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00374722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404238 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20404267 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 939888 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 404251 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 22975 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3401252 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15820 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3401252 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3401252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404219 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404207 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404213 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20404205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00374684 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 126986 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00121285 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P17712294 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35222889 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15213642 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00121283 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15085132 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374689 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374692 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374716 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374653 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374687 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374659 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374648 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374666 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374727 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374697 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374745 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374699 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374709 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374662 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374671 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374665 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374669 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374747 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374683 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374644 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374713 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374723 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374676 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374700 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374695 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374642 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15085147 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374680 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374646 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374705 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374740 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374655 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15085131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374744 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374650 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374726 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374730 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374674 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374750 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374661 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374707 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374657 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374742 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374703 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00374651 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15213642 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003130_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902931 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902933 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7902932 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7902930 | - | - |
| Affymetrix Human Genome U219 Array | 11752992_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11752991_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11752991_at | - | - |
| Affymetrix Human Genome U219 Array | 11752992_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025751 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025751 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 101065 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1657402 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1690894 | NR_003130.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1657402 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1690894 | NR_003130.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690894 | NR_003130.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1657402 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2092516 | NR_003130.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1690894 | NR_003130.2 | human_transcriptome |

HSP90B3P
Genomic Map