Assay Details
Gene Symbol: FRRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005270861.3 | XP_005270918.1 | 15 - 16 | 1756 | 100 | |
XM_011541451.2 | XP_011539753.1 | 15 - 16 | 2229 | 100 | ||
XM_017001271.1 | XP_016856760.1 | 13 - 14 | 1640 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000287474.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000370176.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000489209.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | ENST00000492943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015049.hg.1 | NM_001013660.2 | Refseq |
