Assay Details
Gene Symbol: LOC102606465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC045807.2 | - | 6 - 7 | 884 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
