Assay Details
| Gene Symbol: NRADDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024046.1 | 3 - 4 | 381 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurobeachin like 2 | NBEAL2 |
BDPLT4, GPS |
-2.9kb | Sense | Chr. 3 - 46979683 - 47009704 on NCBI Build GRCh38 |
| Downstream | SET domain containing 2 | SETD2 |
HBP231, HIF-1, HIP-1, HSPC069, HYPB, KMT3A, LLS, SET2, p231HBP |
2.9kb | Anti-sense | Chr. 3 - 47016408 - 47164109 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000264.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 39434 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 51479 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 361554 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3386150 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4449 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4448 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50423 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3386150 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3386150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217143 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217152 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217143 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22351313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02217145 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P103532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217160 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217163 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217155 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217150 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217145 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217158 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217127 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217136 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217147 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217131 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217143 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15434389 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217166 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217139 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15434391 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217129 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217141 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217153 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217124 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217123 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02217134 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P103532 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03005423.hg.1 | TC03000264.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005423.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151657.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151657.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151657.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151657.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151657.hg.1 | TC0300007279.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | ENST00000437305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007279.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151658.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151658.hg.1 | NR_024046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151658.hg.1 | TC0300007279.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022571 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022572 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022572 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022571 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 165153 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3243603 | NR_024046.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3243603 | BX117367.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243603 | NR_024046.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3243603 | BX117367.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243603 | NR_024046.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243603 | BX117367.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243603 | NR_024046.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3243603 | BX117367.1 | human_transcriptome |

NRADDP
Genomic Map