Assay Details
| Gene Symbol: SCN11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005647.1 | XP_016861136.1 | 3 - 4 | 464 | 132 | |
| XM_017005648.1 | XP_016861137.1 | 1 - 2 | 143 | 132 | ||
| XM_017005652.1 | XP_016861141.1 | 1 - 2 | 143 | 132 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017005648.1, XM_017005652.1 transcripts.
Set Membership: |
|

SCN11A
Genomic Map