Assay Details
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110070.1 | 1 - 2 | 41 | 65 | ||
GenBank mRNA | BM544796.1 | - | 1 - 2 | 62 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144561.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700144561.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144561.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144561.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700144561.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144561.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144561.hg.1 | TC0700006493.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000413706.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000422230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | ENST00000423008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110066.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110067.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110068.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700006493.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144557.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144557.hg.1 | NR_110070.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700144557.hg.1 | TC0700006493.hg.1 | Clariom D TC |
