Assay Details
Gene Symbol: TNRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017012738.1 | XP_016868227.1 | 1 - 2 | 124 | 74 | |
GenBank mRNA | BC029940.1 | - | 2 - 3 | 211 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012738.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000328270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000328270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000328270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000328270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000328270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399434.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399434.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399434.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399434.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000399537.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000413081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000413081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000413081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000413081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000413081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000430969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000434361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000434361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000434361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000434361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000440081.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000440081.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000440081.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000440081.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000455076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000455076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000455076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000455076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000455076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000464852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000464852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000464852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000464852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000497663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000497663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | ENST00000497663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010155.hg.1 | NM_001080495.2 | Refseq |
