Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045254.hg.1 |
TC01002908.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045254.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045254.hg.1 |
ENST00000370188.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045254.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045254.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000534652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
ENST00000534652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000534652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
ENST00000534652.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015042.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
ENST00000263177.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
ENST00000370188.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
NM_001010861.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
NM_001037317.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212077.hg.1 |
TC0100015042.hg.1 |
Clariom D TC |