Assay Details
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010861.2 | NP_001010861.1 | 5 - 6 | 1141 | 105 | |
GenBank mRNA | AK295688.1 | - | 5 - 6 | 879 | 105 | |
AY634620.1 | - | 5 - 6 | 974 | 105 | ||
BC040174.1 | - | 5 - 6 | 878 | 105 | ||
BC136350.1 | - | 5 - 6 | 1079 | 9020337 | 105 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002908.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000534652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | ENST00000534652.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045248.hg.1 | TC01002908.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045248.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045248.hg.1 | ENST00000370188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045248.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045248.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045244.hg.1 | TC01002908.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045244.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045244.hg.1 | ENST00000370188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045244.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045244.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045250.hg.1 | TC01002908.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045250.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045250.hg.1 | ENST00000370188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045250.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045250.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045246.hg.1 | TC01002908.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045246.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045246.hg.1 | ENST00000370188.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045246.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045246.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212072.hg.1 | TC0100015042.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212073.hg.1 | TC0100015042.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000263177.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000370188.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000534652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | ENST00000534652.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001010861.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015042.hg.1 | NM_001037317.1 | Refseq |
