Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007686.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155575.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155575.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155575.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155575.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155575.hg.1 |
TC0300007686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155574.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155574.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155574.hg.1 |
ENST00000473434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155574.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155574.hg.1 |
TC0300007686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300155577.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155577.hg.1 |
NR_046702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155577.hg.1 |
TC0300007686.hg.1 |
Clariom D TC |