| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147048.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147048.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147048.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147048.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147048.hg.1 |
TC0300006752.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300147041.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147041.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147041.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147041.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147041.hg.1 |
TC0300006752.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300006752.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147043.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147043.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147043.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147043.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147043.hg.1 |
TC0300006752.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300147052.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147052.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147052.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147052.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147052.hg.1 |
TC0300006752.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300147053.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147053.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147053.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147053.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147053.hg.1 |
TC0300006752.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300147051.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147051.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147051.hg.1 |
ENST00000429949.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300147051.hg.1 |
NR_103843.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300147051.hg.1 |
TC0300006752.hg.1 |
Clariom D TC |