Assay Details
Gene Symbol: TMEM89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008269.1 | NP_001008270.1 | 1 - 2 | 299 | 55 | |
GenBank mRNA | BC133034.1 | - | 1 - 2 | 398 | 40147056 | 55 |
BC133036.1 | - | 1 - 2 | 398 | 40147058 | 55 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001385.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223399 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223382 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223375 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223374 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223388 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223391 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223393 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22357808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223396 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22357811 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22357806 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15435466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02223390 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P121282 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P121282 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 33950 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50943 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P121282 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P121282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223375 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223392 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223377 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223386 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223388 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223393 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223374 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223396 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223390 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223383 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223390 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223396 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223377 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223375 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223386 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223374 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223384 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223388 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223393 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223383 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357806 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223382 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02223392 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P121282 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223388 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223375 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223394 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223393 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223377 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223384 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223390 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223396 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223386 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223382 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P02223398 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P121282 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223369 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223386 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223438 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223388 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223437 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223393 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223445 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223442 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223440 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435466 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223379 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223435 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223429 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223375 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223390 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223447 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223377 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223382 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223384 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02223427 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001008269_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8087118 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8087117 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8087116 | - | - |
Affymetrix Human Genome U219 Array | 11737403_at | - | - |
Affymetrix Human Genome U219 Array | 11737403_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR03024219.hg.1 | TC03001385.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03024219.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03024219.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03024220.hg.1 | TC03001385.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03024220.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03024220.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011030.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011030.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011030.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011030.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011030.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181006.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181006.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181006.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181006.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181006.hg.1 | TC0300011030.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300181004.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181004.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181004.hg.1 | TC0300011030.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300181007.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181007.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181007.hg.1 | TC0300011030.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300181005.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181005.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181005.hg.1 | ENST00000330862.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300181005.hg.1 | NM_001008269.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300181005.hg.1 | TC0300011030.hg.1 | Clariom D TC |
Human Genome U95 Set | 48968_PM_at | - | - |
Human Genome U95 Set | 48968_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021034 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021034 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 114265 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1694339 | BC133036.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_17371-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7320100 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_17371 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1694339 | BC133034.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1694339 | BC133036.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1694339 | NM_001008269.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1694339 | BC133034.1 | human_transcriptome |
