Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026943.hg.1 |
TC07001735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026943.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026943.hg.1 |
ENST00000480514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026943.hg.1 |
ENST00000485614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026943.hg.1 |
ENST00000486180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026943.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026943.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026948.hg.1 |
TC07001735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026948.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026948.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026945.hg.1 |
TC07001735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026945.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026945.hg.1 |
ENST00000480514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026945.hg.1 |
ENST00000485614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026945.hg.1 |
ENST00000486180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026945.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026944.hg.1 |
TC07001735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026944.hg.1 |
ENST00000480514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026944.hg.1 |
ENST00000485614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026944.hg.1 |
ENST00000486180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026944.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026944.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187691.hg.1 |
TC0700012163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187692.hg.1 |
TC0700012163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187693.hg.1 |
TC0700012163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700187696.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187696.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187696.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700187696.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700187696.hg.1 |
TC0700012163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000460135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000469099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000480514.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000485614.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000486180.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
ENST00000490493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012163.hg.1 |
NR_027068.1 |
Refseq |