Assay Details
Gene Symbol: WAC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033805.1 | 1 - 2 | 616 | 79 | ||
GenBank mRNA | BC045818.1 | - | 1 - 2 | 596 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105376468 | LOC105376468 | -89.7kb | Sense | Chr. 10 - 28433008 - 28495813 on NCBI Build GRCh38 | |
Downstream | WW domain containing adaptor with coiled-coil | WAC |
BM-016, DESSH, PRO1741, Wwp4 |
234.0 | Anti-sense | Chr. 10 - 28532426 - 28623112 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001140.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10014294.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014294.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014294.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014294.hg.1 | TC10001140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014296.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014296.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014296.hg.1 | TC10001140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014295.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014295.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014295.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014295.hg.1 | TC10001140.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169166.hg.1 | TC1000010145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169171.hg.1 | TC1000010145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169167.hg.1 | TC1000010145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169168.hg.1 | TC1000010145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000527986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010145.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169175.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169175.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169175.hg.1 | ENST00000528337.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169175.hg.1 | NR_033805.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169175.hg.1 | TC1000010145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000254181.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000598197.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600020.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | ENST00000600940.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008982.hg.1 | NM_020903.2 | Refseq |
