Assay Details
Gene Symbol: HNRNPUL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079559.2 | NP_001073027.1 | 9 - 10 | 1830 | 95 | |
GenBank mRNA | DQ470474.1 | - | 9 - 10 | 1794 | 95 |
Gene Symbol: HNRNPUL2-BSCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037946.1 | 9 - 10 | 1829 | 95 | ||
GenBank mRNA | AK122942.1 | - | 9 - 10 | 1829 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11003488.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK122942.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11021973.hg.1 | TC11003488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11021973.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11021973.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013175.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | ENST00000540127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013176.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204419.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204419.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204419.hg.1 | ENST00000403734.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204419.hg.1 | NR_037946.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204419.hg.1 | TC1100013175.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100204436.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204436.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204436.hg.1 | ENST00000301785.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100204436.hg.1 | NM_001079559.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100204436.hg.1 | TC1100013176.hg.1 | Clariom D TC |
