Assay Details
| Gene Symbol: TECRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010874.4 | NP_001010874.2 | 8 - 9 | 885 | 102 | |
| XM_005265662.4 | XP_005265719.1 | 7 - 8 | 889 | 102 | ||
| XM_005265663.2 | XP_005265720.1 | 8 - 9 | 885 | 102 | ||
| XM_005265664.2 | XP_005265721.1 | 8 - 9 | 885 | 102 | ||
| XM_005265665.3 | XP_005265722.1 | 8 - 9 | 885 | 102 | ||
| GenBank mRNA | AK290606.1 | - | 8 - 9 | 881 | 102 | |
| AL833108.1 | - | 8 - 9 | 886 | 102 | ||
| BX647590.1 | - | 8 - 9 | 822 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001242.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016857.hg.1 | TC04001242.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016857.hg.1 | ENST00000381210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016857.hg.1 | ENST00000507440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016857.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400173731.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400173731.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400173731.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400173731.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400173731.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400173731.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400173731.hg.1 | TC0400010853.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000381210.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000507440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000509536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000511997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | ENST00000513125.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010853.hg.1 | NM_001010874.4 | Refseq |

TECRL
Genomic Map