| Array Name | 
									Probe IDs | 
									Targets | 
									Target Type | 
								
	
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569428.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569428.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569428.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160639.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160639.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160639.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160639.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160639.hg.1 | 
												TC1100008217.hg.1 | 
												Clariom D TC | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160640.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160640.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160640.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160640.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1100160640.hg.1 | 
												TC1100008217.hg.1 | 
												Clariom D TC | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000538407.2 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569428.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569428.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569428.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												ENST00000569432.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1100008217.hg.1 | 
												NR_120541.1 | 
												Refseq |