Assay Details
| Gene Symbol: LINC-PINT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034120.1 | 1 - 2 | 365 | 74 | ||
| NR_110472.1 | 1 - 2 | 365 | 74 | |||
| NR_110473.1 | 1 - 2 | 365 | 74 | |||
| GenBank mRNA | AK095549.1 | - | 1 - 2 | 368 | 74 | |
| GU321462.1 | - | 1 - 2 | 186 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001868.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000385015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_029503.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_029517.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028967.hg.1 | TC07001868.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028967.hg.1 | ENST00000432045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028967.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028968.hg.1 | TC07001868.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028968.hg.1 | ENST00000432045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028968.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028970.hg.1 | TC07001868.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028970.hg.1 | ENST00000432045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07028970.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190965.hg.1 | TC0700012613.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190968.hg.1 | TC0700012613.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700190964.hg.1 | TC0700012613.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000362111.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000385015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000418546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000432045.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | ENST00000447307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_029503.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_029517.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_034120.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012613.hg.1 | NR_110473.1 | Refseq |

LINC-PINT
Genomic Map