Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000156.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000156.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000156.hg.1 |
TC10000012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000157.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000157.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000157.hg.1 |
TC10000012.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000144347.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144347.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144347.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144347.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144347.hg.1 |
TC1000006469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000144350.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144350.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144350.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144350.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144350.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144350.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144350.hg.1 |
TC1000006469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000144349.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144349.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144349.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144349.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144349.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000144349.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000144349.hg.1 |
TC1000006469.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000425630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
ENST00000583117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006469.hg.1 |
NR_015376.2 |
Refseq |