Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010093.hg.1 |
TC07000649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010093.hg.1 |
ENST00000313669.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010093.hg.1 |
ENST00000397927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010093.hg.1 |
ENST00000528707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010093.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160787.hg.1 |
TC0700008592.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000313669.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
ENST00000613501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_001278563.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008592.hg.1 |
NM_133457.4 |
Refseq |