Assay Details
| Gene Symbol: GGPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB016043.1 | - | 1 - 2 | 144 | 80 | |
| AB019036.1 | - | 1 - 2 | 185 | 80 | ||
| AF057698.1 | - | 1 - 2 | 213 | 80 | ||
| AF125394.1 | - | 1 - 2 | 122 | 80 | ||
| AK025139.1 | - | 1 - 2 | 207 | 80 | ||
| BC005252.1 | - | 1 - 2 | 207 | 4051481 | 80 | |
| BI462253.1 | - | 1 - 2 | 234 | 80 | ||
| CB216084.1 | - | 1 - 2 | 225 | 80 | ||
| DB473584.1 | - | 1 - 2 | 235 | 80 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB016043.1,AB019036.1,AF057698.1,AF125394.1,BC005252.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000254724.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000254724.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000358966.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000471812.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000488594.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | ENST00000497327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NM_001037277.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012055.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000282841.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000391855.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000482013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | ENST00000489692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | NR_036605.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188819.hg.1 | TC0100012055.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 130606 | - | - |

GGPS1
Genomic Map