Assay Details
| Gene Symbol: NRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244972.1 | NP_001231901.1 | 12 - 13 | 2421 | 75 | |
| NM_001244973.1 | NP_001231902.1 | 12 - 13 | 2421 | 75 | ||
| NM_003873.5 | NP_003864.4 | 12 - 13 | 2442 | 75 | ||
| NR_045259.1 | 11 - 12 | 2239 | 75 | |||
| XM_006717521.1 | XP_006717584.1 | 12 - 13 | 2216 | 75 | ||
| XM_006717522.1 | XP_006717585.1 | 12 - 13 | 2216 | 75 | ||
| XM_006717523.2 | XP_006717586.1 | 12 - 13 | 2216 | 75 | ||
| XM_006717524.1 | XP_006717587.1 | 11 - 12 | 2111 | 75 | ||
| XM_006717525.1 | XP_006717588.1 | 11 - 12 | 2111 | 75 | ||
| XM_017016865.1 | XP_016872354.1 | 12 - 13 | 2195 | 75 | ||
| XM_017016866.1 | XP_016872355.1 | 11 - 12 | 2036 | 75 | ||
| GenBank mRNA | AB209641.1 | - | 12 - 13 | 2070 | 75 | |
| AF016050.1 | - | 12 - 13 | 1919 | 75 | ||
| AF018956.1 | - | 12 - 13 | 1919 | 75 | ||
| AK292822.1 | - | 12 - 13 | 2087 | 75 | ||
| AL833608.1 | - | 11 - 12 | 1970 | 75 | ||
| AY249243.1 | - | 12 - 13 | 1919 | 75 | ||
| BX510902.1 | - | 12 - 13 | 2106 | 75 | ||
| BX648250.1 | - | 12 - 13 | 2390 | 75 | ||
| CR749333.1 | - | 12 - 13 | 2443 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001166.hg.1
Set Membership: |
|

NRP1
Genomic Map