Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004009.hg.1 |
TC07000261.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004009.hg.1 |
ENST00000265522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004009.hg.1 |
ENST00000395891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004009.hg.1 |
ENST00000453890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004009.hg.1 |
NM_015052.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
ENST00000395891.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
ENST00000453890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
NM_001287059.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
NM_015052.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
ENST00000395891.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
ENST00000453890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
NM_001287059.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
NM_015052.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700150868.hg.1 |
TC0700007336.hg.1 |
Clariom D TC |