Assay Details
| Gene Symbol: MIOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012361.1 | XP_016867850.1 | 1 - 2 | 386 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012361.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000461907.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000461907.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000461907.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000340080.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000405785.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000433635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000445169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000456533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000461907.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000461907.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000461907.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000479694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | ENST00000493227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700006669.hg.1 | NM_019005.3 | Refseq |

MIOS
Genomic Map