Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013536.hg.1 |
TC0X001020.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013536.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X013536.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000442430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000442430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000442430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000442430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000442430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000442430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
ENST00000485431.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009637.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165661.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165661.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165661.hg.1 |
ENST00000376509.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00165661.hg.1 |
NM_006875.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00165661.hg.1 |
TC0X00009637.hg.1 |
Clariom D TC |