Assay Details
| Gene Symbol: CCDC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014008.4 | NP_054727.1 | 8 - 9 | 1144 | 85 | |
| XM_005272599.3 | XP_005272656.1 | 8 - 9 | 1073 | 85 | ||
| GenBank mRNA | AJ005890.1 | - | 8 - 9 | 1060 | 85 | |
| AK291976.1 | - | 8 - 9 | 1105 | 85 | ||
| AK298264.1 | - | 8 - 9 | 1139 | 85 | ||
| AK316375.1 | - | 8 - 9 | 1090 | 85 | ||
| BC000972.2 | - | 8 - 9 | 1091 | 3449051 | 85 | |
| BC011675.2 | - | 8 - 9 | 975 | 4299954 | 85 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000269.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cellular copper ion homeostasis
Golgi to plasma membrane transport
cytoplasmic sequestering of NF-kappaB
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein binding
cullin family protein binding
cellular copper ion homeostasis
Golgi to plasma membrane transport
cytoplasmic sequestering of NF-kappaB
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein binding
cullin family protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003505.hg.1 | TC0X000269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003505.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003505.hg.1 | ENST00000538876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003505.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003504.hg.1 | TC0X000269.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003504.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003504.hg.1 | ENST00000538876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X003504.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149573.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149573.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149573.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00149573.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00149573.hg.1 | TC0X00007224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |

CCDC22
Genomic Map