Assay Details
| Gene Symbol: KPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291296.1 | NP_001278225.1 | 4 - 5 | 534 | 129 | |
| NM_007059.3 | NP_008990.2 | 6 - 7 | 702 | 129 | ||
| XM_011526399.2 | XP_011524701.1 | 9 - 10 | 2207 | 129 | ||
| XM_017026226.1 | XP_016881715.1 | 9 - 10 | 1052 | 129 | ||
| XM_017026227.1 | XP_016881716.1 | 8 - 9 | 1128 | 129 | ||
| XM_017026228.1 | XP_016881717.1 | 9 - 10 | 1052 | 129 | ||
| GenBank mRNA | AF105369.1 | - | 6 - 7 | 634 | 129 | |
| AK023996.1 | - | 6 - 7 | 657 | 129 | ||
| AK225419.1 | - | 6 - 7 | 646 | 129 | ||
| BC009249.2 | - | 6 - 7 | 617 | 3029623 | 129 | |
| DR759729.1 | - | 4 - 5 | 446 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

KPTN
Genomic Map