Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009043.hg.1 |
ENST00000309424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009043.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009043.hg.1 |
TC19000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000590794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009040.hg.1 |
ENST00000309424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009040.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009040.hg.1 |
TC19000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009045.hg.1 |
ENST00000309424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009045.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009045.hg.1 |
TC19000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009041.hg.1 |
ENST00000309424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009041.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009041.hg.1 |
TC19000646.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
TC1900008323.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163861.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163853.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163853.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163853.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163853.hg.1 |
TC1900008323.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900163853.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163853.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163853.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000589804.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000590794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008323.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163852.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163852.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163852.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163852.hg.1 |
TC1900008323.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900163852.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163852.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163852.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
NM_012099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
TC1900008323.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
ENST00000309424.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
ENST00000592852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
NM_001297590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900163854.hg.1 |
NM_012099.1 |
Refseq |