Assay Details
Gene Symbol: CD3EAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297590.1 | NP_001284519.1 | 2 - 3 | 656 | 61 | |
NM_012099.1 | NP_036231.1 | 2 - 3 | 650 | 61 | ||
XM_017026180.1 | XP_016881669.1 | 2 - 3 | 751 | 61 | ||
GenBank mRNA | AF017633.1 | - | 2 - 3 | 174 | 61 | |
AK292183.1 | - | 2 - 3 | 215 | 61 | ||
BC009705.1 | - | 2 - 3 | 207 | 61 | ||
BC038992.1 | - | 2 - 3 | 178 | 6052122 | 61 | |
BC046141.1 | - | 2 - 3 | 450 | 61 | ||
BC054044.1 | - | 2 - 3 | 526 | 61 | ||
BC065251.1 | - | 2 - 3 | 181 | 61 | ||
BC108889.2 | - | 2 - 3 | 415 | 40001010 | 61 | |
BU166593.1 | - | 2 - 3 | 525 | 61 | ||
U86751.1 | - | 2 - 3 | 650 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017026180.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
