Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002816.hg.1 |
ENST00000361462.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002816.hg.1 |
ENST00000361577.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002816.hg.1 |
ENST00000382233.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002816.hg.1 |
NM_015091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14002816.hg.1 |
TC14000262.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
ENST00000361462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
ENST00000361577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
ENST00000557423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
NM_001308120.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
NM_015091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
NR_131765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
ENST00000361462.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
ENST00000361577.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
ENST00000557423.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
NM_001308120.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
NM_015091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
NR_131765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400148847.hg.1 |
TC1400007033.hg.1 |
Clariom D TC |