Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018229.hg.1 |
TC0X001359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018229.hg.1 |
ENST00000370978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018229.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018230.hg.1 |
TC0X001359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018230.hg.1 |
ENST00000066465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018230.hg.1 |
ENST00000370978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018230.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173310.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173310.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173310.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173310.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173310.hg.1 |
TC0X00010804.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173311.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173311.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173311.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173311.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173311.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173311.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173311.hg.1 |
TC0X00010804.hg.1 |
Clariom D TC |