Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018240.hg.1 |
TC0X001359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018240.hg.1 |
ENST00000066465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018240.hg.1 |
ENST00000370978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018240.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018242.hg.1 |
TC0X001359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018242.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018239.hg.1 |
TC0X001359.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018239.hg.1 |
ENST00000066465.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018239.hg.1 |
ENST00000370978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018239.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173320.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173320.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173320.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173320.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173320.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173320.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173320.hg.1 |
TC0X00010804.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010804.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173322.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173322.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173322.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173322.hg.1 |
ENST00000370978.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173322.hg.1 |
ENST00000447817.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00173322.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173322.hg.1 |
TC0X00010804.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00173324.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173324.hg.1 |
NM_017666.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00173324.hg.1 |
TC0X00010804.hg.1 |
Clariom D TC |