| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008270.hg.1 |
TC04000566.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008270.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008270.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008269.hg.1 |
TC04000566.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008269.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008269.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400157674.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400157674.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400157674.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400157674.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400157674.hg.1 |
TC0400008389.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400157673.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400157673.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400157673.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400157673.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400157673.hg.1 |
TC0400008389.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
ENST00000317735.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008389.hg.1 |
NM_006583.2 |
Refseq |