Assay Details
| Gene Symbol: ACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005722.3 | NP_005713.1 | 2 - 3 | 369 | 150 | |
| GenBank mRNA | AF006082.1 | - | 2 - 3 | 228 | 150 | |
| AK315205.1 | - | 2 - 3 | 218 | 150 | ||
| BC014546.1 | - | 2 - 3 | 235 | 4247021 | 150 | |
| BC036253.1 | - | 2 - 3 | 377 | 5268748 | 150 | |
| BI670123.1 | - | 2 - 3 | 306 | 150 | ||
| DC404298.1 | - | 2 - 3 | 369 | 150 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000378.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC036253.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
establishment or maintenance of cell polarity
asymmetric cell division
meiotic chromosome movement towards spindle pole
cytosolic transport
meiotic cytokinesis
Arp2/3 complex-mediated actin nucleation
Fc-gamma receptor signaling pathway involved in phagocytosis
cilium assembly
ephrin receptor signaling pathway
spindle localization
structural constituent of cytoskeleton
ATP binding
actin filament binding
movement of cell or subcellular component
establishment or maintenance of cell polarity
asymmetric cell division
meiotic chromosome movement towards spindle pole
cytosolic transport
meiotic cytokinesis
Arp2/3 complex-mediated actin nucleation
Fc-gamma receptor signaling pathway involved in phagocytosis
cilium assembly
ephrin receptor signaling pathway
spindle localization
structural constituent of cytoskeleton
ATP binding
actin filament binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | TC02000378.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | ENST00000260641.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | ENST00000377982.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | ENST00000476840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | ENST00000535303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | ENST00000542850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005734.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007469.hg.1 | ENST00000521432.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007469.hg.1 | ENST00000521432.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200154051.hg.1 | TC0200007835.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000260641.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000377982.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000471552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000476840.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | ENST00000542850.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_001005386.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007835.hg.1 | NM_005722.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007469.hg.1 | ENST00000521432.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007469.hg.1 | ENST00000521432.2 | Ensembl |

ACTR2
Genomic Map