Assay Details
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005268012.2 | XP_005268069.1 | 1 - 2 | 233 | 66 | |
GenBank mRNA | BG717628.1 | - | 1 - 2 | 241 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005268012.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
