Assay Details
| Gene Symbol: GNPNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005268012.2 | XP_005268069.1 | 2 - 3 | 264 | 127 | |
| GenBank mRNA | BG717628.1 | - | 2 - 3 | 272 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005268012.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011639.hg.1 | ENST00000604710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000216410.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000553987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000554421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | ENST00000557604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009190.hg.1 | NM_198066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011639.hg.1 | ENST00000604710.1 | Ensembl |

GNPNAT1
Genomic Map