Assay Details
Gene Symbol: CCNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004354.2 | NP_004345.1 | 2 - 3 | 442 | 113 | |
XM_011532398.1 | XP_011530700.1 | 2 - 3 | 163 | 113 | ||
XM_011532399.2 | XP_011530701.1 | 2 - 3 | 582 | 113 | ||
GenBank mRNA | AK292029.1 | - | 3 - 4 | 303 | 113 | |
AK293899.1 | - | 1 - 2 | 246 | 113 | ||
BC032518.1 | - | 2 - 3 | 431 | 5494024 | 113 | |
BT019503.1 | - | 1 - 2 | 139 | 113 | ||
CR542181.1 | - | 1 - 2 | 139 | 113 | ||
CR542200.1 | - | 1 - 2 | 139 | 113 | ||
DC344594.1 | - | 2 - 3 | 442 | 113 | ||
L49506.1 | - | 1 - 2 | 375 | 113 | ||
U47414.1 | - | 2 - 3 | 274 | 113 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000431.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | TC04000431.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | ENST00000316355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | ENST00000354403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | ENST00000395640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | ENST00000497512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | ENST00000502280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006098.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188533.hg.1 | TC0400012818.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
