Assay Details
Gene Symbol: CCNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004354.2 | NP_004345.1 | 4 - 5 | 832 | 128 | |
XM_011532398.1 | XP_011530700.1 | 4 - 5 | 553 | 128 | ||
XM_011532399.2 | XP_011530701.1 | 4 - 5 | 972 | 128 | ||
GenBank mRNA | AK292029.1 | - | 5 - 6 | 693 | 128 | |
AK293899.1 | - | 3 - 4 | 636 | 128 | ||
BC032518.1 | - | 4 - 5 | 821 | 5494024 | 128 | |
BT019503.1 | - | 3 - 4 | 529 | 128 | ||
CR542181.1 | - | 3 - 4 | 529 | 128 | ||
CR542200.1 | - | 3 - 4 | 529 | 128 | ||
L49506.1 | - | 3 - 4 | 765 | 128 | ||
U47414.1 | - | 4 - 5 | 664 | 128 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000431.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | TC04000431.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | ENST00000316355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | ENST00000354403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | ENST00000395640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | ENST00000497512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | ENST00000502280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006100.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | TC04000431.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | ENST00000316355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | ENST00000354403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | ENST00000395640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | ENST00000497512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | ENST00000502280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006101.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188536.hg.1 | TC0400012818.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188538.hg.1 | TC0400012818.hg.1 | Clariom D TC |
