Assay Details
Gene Symbol: CCNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004354.2 | NP_004345.1 | 6 - 7 | 1015 | 140 | |
XM_011532398.1 | XP_011530700.1 | 6 - 7 | 736 | 140 | ||
XM_011532399.2 | XP_011530701.1 | 6 - 7 | 1155 | 140 | ||
GenBank mRNA | AK292029.1 | - | 7 - 8 | 876 | 140 | |
AK293899.1 | - | 5 - 6 | 819 | 140 | ||
BC032518.1 | - | 6 - 7 | 1004 | 5494024 | 140 | |
BT019503.1 | - | 5 - 6 | 712 | 140 | ||
CR542181.1 | - | 5 - 6 | 712 | 140 | ||
CR542200.1 | - | 5 - 6 | 712 | 140 | ||
L49506.1 | - | 5 - 6 | 948 | 140 | ||
U47414.1 | - | 6 - 7 | 847 | 140 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000431.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | TC04000431.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | ENST00000316355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | ENST00000354403.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | ENST00000395640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | ENST00000497512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | ENST00000502280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04006103.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | ENST00000512918.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012818.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012819.hg.1 | ENST00000514756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000316355.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000354403.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000395640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000502280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | ENST00000509972.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | NM_004354.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400188542.hg.1 | TC0400012818.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 109215 | - | - |
