Assay Details
Gene Symbol: GPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142466.2 | NP_001135938.1 | 9 - 10 | 1292 | 117 | |
NM_133443.3 | NP_597700.1 | 9 - 10 | 1316 | 117 | ||
XM_017023790.1 | XP_016879279.1 | 6 - 7 | 789 | 117 | ||
GenBank mRNA | AK094971.1 | - | 9 - 10 | 1216 | 117 | |
AY029173.2 | - | 9 - 10 | 1300 | 117 | ||
BC062555.1 | - | 9 - 10 | 1265 | 6429257 | 117 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000426.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16005895.hg.1 | ENST00000340124.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16005895.hg.1 | ENST00000440783.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16005895.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16005895.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16005895.hg.1 | TC16000426.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600156361.hg.1 | TC1600007723.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
