Assay Details
Gene Symbol: KCNQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004518.5 | NP_004509.2 | 9 - 10 | 1402 | 130 | |
XM_011528811.1 | XP_011527113.1 | 9 - 10 | 1314 | 130 | ||
GenBank mRNA | Y15065.1 | - | 9 - 10 | 1230 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
